siRNA Details
| Gene Symbol: Krt40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039666.1 | NP_001034755.1 | 4 | 778 | ||
| GenBank mRNA | BC107359.1 | - | 4 | 778 | ||
| BC107360.2 | - | 4 | 776 | |||
| BC119522.2 | - | 4 | 778 | 40055959 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt40| Gene Symbol: Krt40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039666.1 | NP_001034755.1 | 4 | 778 | ||
| GenBank mRNA | BC107359.1 | - | 4 | 778 | ||
| BC107360.2 | - | 4 | 776 | |||
| BC119522.2 | - | 4 | 778 | 40055959 | ||