siRNA Details
| Gene Symbol: Znrf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080924.2 | NP_001074393.1 | 8 | 2298 | ||
| NM_001290501.1 | NP_001277430.1 | 9 | 2145 | |||
| GenBank mRNA | AK133342.1 | - | 9 | 2146 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znrf3| Gene Symbol: Znrf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080924.2 | NP_001074393.1 | 8 | 2298 | ||
| NM_001290501.1 | NP_001277430.1 | 9 | 2145 | |||
| GenBank mRNA | AK133342.1 | - | 9 | 2146 | ||