siRNA Details
| Gene Symbol: Znrf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080924.2 | NP_001074393.1 | 2 | 509 | ||
| NM_001290501.1 | NP_001277430.1 | 3 | 356 | |||
| GenBank mRNA | AK133342.1 | - | 3 | 357 | ||
| AK163639.1 | - | 2 | 509 | |||
| BC151080.1 | - | 3 | 230 | 9087993 | ||
| BC151083.1 | - | 3 | 230 | 9087996 | ||

Znrf3
Genomic Map