siRNA Details
| Gene Symbol: F630007L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK154187.1 | - | 2248 | |||
| Gene Symbol: BC005512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135827.1 | - | 3 | 2190 | ||
| AV303415.2 | - | 115 | ||||
| AW215732.1 | - | 136 | ||||
| Gene Symbol: Gm29673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC002257.1 | - | 1 | 1003 | ||
| Gene Symbol: Gm6958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK149728.1 | - | 1 | 1304 | ||

F630007L15Rik
Genomic Map