siRNA Details
| Gene Symbol: Nkain2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013411.2 | NP_001013429.2 | 3 | 450 | ||
| NM_001025286.2 | NP_001020457.2 | 3 | 450 | |||
| GenBank mRNA | AY940478.1 | - | 3 | 246 | ||
| BC141063.1 | - | 3 | 320 | 9055675 | ||
| DQ065606.1 | - | 3 | 383 | |||
| DQ065607.1 | - | 3 | 383 | |||
| EF058049.2 | - | 3 | 383 | |||
| EF058050.1 | - | 2 | 231 | |||
| EF058051.1 | - | 3 | 231 | |||

Nkain2
Genomic Map