siRNA Details
| Gene Symbol: Trdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029726.2 | NP_084002.2 | 31, 32 | 1958 | ||
| GenBank mRNA | AK036470.1 | - | 13, 14 | 498 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trdn| Gene Symbol: Trdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029726.2 | NP_084002.2 | 31, 32 | 1958 | ||
| GenBank mRNA | AK036470.1 | - | 13, 14 | 498 | ||