siRNA Details
| Gene Symbol: Gnptab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004164.2 | NP_001004164.2 | 5 | 467 | ||
| GenBank mRNA | AK140867.1 | - | 4 | 571 | ||
| AK173132.1 | - | 5 | 467 | |||
| BC060638.1 | - | 6 | 575 | 6849405 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnptab| Gene Symbol: Gnptab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004164.2 | NP_001004164.2 | 5 | 467 | ||
| GenBank mRNA | AK140867.1 | - | 4 | 571 | ||
| AK173132.1 | - | 5 | 467 | |||
| BC060638.1 | - | 6 | 575 | 6849405 | ||