siRNA Details
| Gene Symbol: Adcy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009622.1 | NP_033752.1 | 12 | 2158 | ||
| GenBank mRNA | AF053980.1 | - | 12 | 1751 | ||
| AK220469.1 | - | 6 | 629 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adcy1| Gene Symbol: Adcy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009622.1 | NP_033752.1 | 12 | 2158 | ||
| GenBank mRNA | AF053980.1 | - | 12 | 1751 | ||
| AK220469.1 | - | 6 | 629 | |||