siRNA Details
| Gene Symbol: Gm11541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007584.2 | NP_001007585.1 | 2 | 500 | ||
| GenBank mRNA | AK082500.1 | - | 2 | 501 | ||
| BC147833.1 | - | 2 | 501 | 9087550 | ||
| BC147838.1 | - | 2 | 501 | 9087555 | ||

Gm11541
Genomic Map