siRNA Details
| Gene Symbol: Fbxo47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081435.1 | NP_001074904.1 | 8 | 1091 | ||
| GenBank mRNA | BC158049.1 | - | 8 | 1026 | 9007353 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo47| Gene Symbol: Fbxo47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081435.1 | NP_001074904.1 | 8 | 1091 | ||
| GenBank mRNA | BC158049.1 | - | 8 | 1026 | 9007353 | |