siRNA Details
| Gene Symbol: Kif19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102615.1 | NP_001096085.1 | 12 | 1569 | ||
| GenBank mRNA | AK155544.1 | - | 12 | 1533 | ||
| BC151045.1 | - | 12 | 1531 | 9087958 | ||
| BC151053.1 | - | 12 | 1531 | 9087966 | ||
| EF570784.1 | - | 12 | 1444 | |||

Kif19a
Genomic Map