siRNA Details
Gene Symbol: Lrrc14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033042.3 | NP_001028214.1 | 2 | 966 | ||
GenBank mRNA | AK146537.1 | - | 2 | 966 | ||
BC064819.1 | - | 2 | 971 | |||
BC139119.1 | - | 2 | 945 | 8862141 | ||
BC139120.1 | - | 2 | 945 | 8862142 |
Gene Symbol: Ccdc127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168658.1 | NP_001162129.1 | 4 | 1339 | ||
GenBank mRNA | AK164539.1 | - | 4 | 1343 |
