siRNA Details
| Gene Symbol: Lrrc14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033042.3 | NP_001028214.1 | 1 | 826 | ||
| GenBank mRNA | AK146537.1 | - | 1 | 826 | ||
| BC064819.1 | - | 1 | 831 | |||
| BC139119.1 | - | 1 | 805 | 8862141 | ||
| BC139120.1 | - | 1 | 805 | 8862142 | ||
| Gene Symbol: Ccdc127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168658.1 | NP_001162129.1 | 4 | 3152 | ||

Lrrc14b
Genomic Map