siRNA Details
Gene Symbol: Csmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081391.2 | NP_001074860.2 | 5 | 977 | ||
GenBank mRNA | AK140264.1 | - | 5 | 978 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Csmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081391.2 | NP_001074860.2 | 5 | 977 | ||
GenBank mRNA | AK140264.1 | - | 5 | 978 |