siRNA Details
| Gene Symbol: Gabrr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081190.1 | NP_001074659.1 | 6 | 648 | ||
| GenBank mRNA | AK141642.1 | - | 2 | 73 | ||
| BC151105.1 | - | 6 | 698 | 9088018 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabrr3| Gene Symbol: Gabrr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081190.1 | NP_001074659.1 | 6 | 648 | ||
| GenBank mRNA | AK141642.1 | - | 2 | 73 | ||
| BC151105.1 | - | 6 | 698 | 9088018 | ||