siRNA Details
| Gene Symbol: Rmnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025343.5 | NP_079619.4 | 3 | 435 | ||
| GenBank mRNA | AK007323.1 | - | 3 | 383 | ||
| AK075603.1 | - | 3 | 418 | |||
| AK169025.1 | - | 3 | 428 | |||
| BC027299.1 | - | 3 | 383 | 3987394 | ||
| BC036150.1 | - | 2 | 355 | 4977394 | ||
| BY740212.1 | - | 3 | 428 | |||

Rmnd1
Genomic Map