siRNA Details
| Gene Symbol: Acsl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027976.2 | NP_082252.1 | 11 | 1745 | ||
| GenBank mRNA | AK034942.1 | - | 1745 | |||
| AK145894.1 | - | 11 | 1406 | |||
| AK150542.1 | - | 11 | 1411 | |||
| AK150663.1 | - | 11 | 1411 | |||
| AK151378.1 | - | 11 | 1200 | |||
| AK161024.1 | - | 11 | 1098 | |||
| AK219510.1 | - | 4 | 212 | |||
| BC031544.1 | - | 11 | 1179 | 3987201 | ||

Acsl5
Genomic Map