siRNA Details
| Gene Symbol: Acsl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027976.2 | NP_082252.1 | 13 | 1933 | ||
| GenBank mRNA | AK006541.1 | - | 2 | 410 | ||
| AK034942.1 | - | 1933 | ||||
| AK145894.1 | - | 13 | 1594 | |||
| AK150542.1 | - | 13 | 1599 | |||
| AK150663.1 | - | 13 | 1599 | |||
| AK151378.1 | - | 13 | 1388 | |||
| AK161024.1 | - | 13 | 1286 | |||
| AK218456.1 | - | 2 | 106 | |||
| BC031544.1 | - | 13 | 1367 | 3987201 | ||

Acsl5
Genomic Map