siRNA Details
| Gene Symbol: Slc25a36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138756.4 | NP_620095.1 | 5 | 513 | ||
| GenBank mRNA | AK042075.1 | - | 5 | 602 | ||
| AK053646.1 | - | 5 | 629 | |||
| AK078779.1 | - | 5 | 514 | |||
| BC008171.1 | - | 5 | 505 | 3710468 | ||

Slc25a36
Genomic Map