siRNA Details
| Gene Symbol: A930004D18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020807.1 | - | 1 | 118 | ||
| AK044329.1 | - | 1 | 111 | |||
| AK044517.1 | - | 1 | 96 | |||
| EL956282.1 | - | 1 | 118 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930004D18Rik| Gene Symbol: A930004D18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020807.1 | - | 1 | 118 | ||
| AK044329.1 | - | 1 | 111 | |||
| AK044517.1 | - | 1 | 96 | |||
| EL956282.1 | - | 1 | 118 | |||