siRNA Details
| Gene Symbol: Gm17762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011601.1 | - | 1 | 404 | ||
| AK033161.1 | - | 1 | 852 | |||
| AK171729.1 | - | 1 | 542 | |||
| Gene Symbol: A930004D18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044329.1 | - | 2 | 308 | ||

Gm17762
Genomic Map