siRNA Details
| Gene Symbol: Ankrd13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013806.1 | NP_001013828.1 | 9, 10 | 1284 | ||
| GenBank mRNA | AK135906.1 | - | 10, 11 | 1020 | ||
| AK157542.1 | - | 1466 | ||||
| BC065088.1 | - | 8, 9 | 3478 | |||
| BC138991.1 | - | 9, 10 | 1292 | 8862013 | ||
| BC138992.1 | - | 10, 11 | 1271 | 8862014 | ||
| CF736962.1 | - | 2, 3 | 280 | |||

Ankrd13c
Genomic Map