siRNA Details
| Gene Symbol: Ube2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009455.3 | NP_033481.1 | 4 | 421 | ||
| GenBank mRNA | AK090177.1 | - | 4 | 422 | ||
| AK148266.1 | - | 4 | 352 | |||
| BC003781.1 | - | 4 | 338 | 3484294 | ||
| X92665.1 | - | 4 | 383 | |||

Ube2e1
Genomic Map