siRNA Details
| Gene Symbol: Rnf207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033489.2 | NP_001028661.2 | 9, 10 | 1113 | ||
| GenBank mRNA | AK042379.1 | - | 9, 10 | 1096 | ||
| BC023223.1 | - | 9, 10 | 1166 | |||
| BC141256.1 | - | 9, 10 | 997 | 9055868 | ||
| BC141257.1 | - | 9, 10 | 997 | 9055869 | ||

Rnf207
Genomic Map