siRNA Details
| Gene Symbol: Uspl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013378.2 | NP_001013396.2 | 1 | 229 | ||
| NM_001115149.1 | NP_001108621.1 | 1 | 229 | |||
| NM_001115150.1 | NP_001108622.1 | 1 | 229 | |||
| NM_001115151.1 | NP_001108623.1 | 1 | 229 | |||
| NM_001115153.1 | NP_001108625.1 | 1 | 229 | |||
| NM_001286824.1 | NP_001273753.1 | 1 | 229 | |||
| NM_001286825.1 | NP_001273754.1 | 1 | 229 | |||
| GenBank mRNA | AK050969.1 | - | 1 | 213 | ||
| AK088787.1 | - | 1 | 95 | |||
| AK145409.1 | - | 1 | 127 | |||
| AK146820.1 | - | 1 | 112 | |||
| AK161654.1 | - | 1 | 76 | |||
| AK168702.1 | - | 1 | 112 | |||
| AK168981.1 | - | 1 | 123 | |||
| AK179867.1 | - | 1 | 65 | |||
| BB655282.1 | - | 1 | 208 | |||
| BC086664.1 | - | 1 | 71 | |||
| BY203045.1 | - | 1 | 230 | |||
| BY298649.1 | - | 1 | 126 | |||

Uspl1
Genomic Map