siRNA Details
| Gene Symbol: 5830473C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252661.1 | NP_001239590.1 | 9 | 1142 | ||
| GenBank mRNA | CB521232.1 | - | 6 | 723 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5830473C10Rik| Gene Symbol: 5830473C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252661.1 | NP_001239590.1 | 9 | 1142 | ||
| GenBank mRNA | CB521232.1 | - | 6 | 723 | ||