siRNA Details
| Gene Symbol: 4930533I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029757.1 | - | 7 | 974 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930533I22Rik| Gene Symbol: 4930533I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029757.1 | - | 7 | 974 | ||