siRNA Details
| Gene Symbol: Zfp191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021559.2 | NP_067534.2 | 2 | 155 | ||
| GenBank mRNA | AB221566.1 | - | 1 | 14 | ||
| AF149093.1 | - | 1 | 14 | |||
| AK146726.1 | - | 2 | 153 | |||
| AY832930.1 | - | 2 | 194 | |||
| BC011345.1 | - | 2 | 198 | 3992743 | ||
| BC054832.1 | - | 2 | 155 | 6467818 | ||

Zfp191
Genomic Map