siRNA Details
| Gene Symbol: Adgrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144045.1 | - | 5 | 816 | ||
| BC132476.1 | - | 5 | 664 | 40130753 | ||
| BC132480.1 | - | 5 | 664 | 40130757 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AX794837.1 | - | 263 | Chr. - - [-] on Build | ||

Adgrg4
Genomic Map