siRNA Details
| Gene Symbol: Tmprss11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030297.2 | NP_001025468.1 | 5 | 496 | ||
| GenBank mRNA | AM260217.1 | - | 5 | 496 | ||
| BC147157.1 | - | 5 | 477 | 9056205 | ||
| BC147158.1 | - | 5 | 477 | 9056165 | ||

Tmprss11c
Genomic Map