siRNA Details
Gene Symbol: Lrch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081255.1 | NP_001074724.1 | 6 | 889 | ||
GenBank mRNA | AK076529.1 | - | 6 | 879 | ||
AK133482.1 | - | 6 | 867 | |||
AK155550.1 | - | 6 | 887 | |||
BC151058.1 | - | 6 | 867 | 9087971 | ||
BC157924.1 | - | 6 | 867 | 9088097 | ||
BC172144.1 | - | 6 | 867 | 9054814 |
