siRNA Details
Gene Symbol: Gm5134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198635.3 | NP_941037.2 | 14 | 1949 | ||
GenBank mRNA | AK133270.1 | - | 14 | 1950 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198635.3 | NP_941037.2 | 14 | 1949 | ||
GenBank mRNA | AK133270.1 | - | 14 | 1950 |