siRNA Details
| Gene Symbol: Ccin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002787.2 | NP_001002787.1 | 1 | 782 | ||
| GenBank mRNA | AK030197.1 | - | 1 | 785 | ||
| BC150949.1 | - | 1 | 769 | 9087860 | ||
| BC150952.1 | - | 1 | 769 | 9087863 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccin| Gene Symbol: Ccin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002787.2 | NP_001002787.1 | 1 | 782 | ||
| GenBank mRNA | AK030197.1 | - | 1 | 785 | ||
| BC150949.1 | - | 1 | 769 | 9087860 | ||
| BC150952.1 | - | 1 | 769 | 9087863 | ||