siRNA Details
| Gene Symbol: Znhit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005223.2 | NP_001005223.1 | 5 | 457 | ||
| GenBank mRNA | AK002888.1 | - | 5 | 449 | ||
| AK003721.1 | - | 5 | 456 | |||
| AK004259.1 | - | 5 | 459 | |||
| AK014350.1 | - | 5 | 468 | |||
| BC089554.1 | - | 5 | 545 | 30300471 | ||

Znhit3
Genomic Map