siRNA Details
| Gene Symbol: Xkr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011874.1 | NP_001011874.1 | 3 | 1395 | ||
| GenBank mRNA | AY534250.1 | - | 3 | 1395 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xkr4| Gene Symbol: Xkr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011874.1 | NP_001011874.1 | 3 | 1395 | ||
| GenBank mRNA | AY534250.1 | - | 3 | 1395 | ||