siRNA Details
| Gene Symbol: Hspa14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015765.2 | NP_056580.2 | 9 | 891 | ||
| GenBank mRNA | AK031489.1 | - | 830 | |||
| AK145937.1 | - | 9 | 845 | |||
| AK153397.1 | - | 9 | 853 | |||
| BC002056.1 | - | 9 | 819 | 3493119 | ||
| BU505607.1 | - | 6 | 542 | |||
| U08215.1 | - | 9 | 851 | |||

Hspa14
Genomic Map