siRNA Details
| Gene Symbol: Ercc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015769.2 | NP_056584.2 | 11 | 2919 | ||
| GenBank mRNA | AF189285.1 | - | 11 | 2919 | ||
| BC026792.1 | - | 11 | 2710 | 4911382 | ||
| BC051036.1 | - | 10 | 2738 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ercc4| Gene Symbol: Ercc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015769.2 | NP_056584.2 | 11 | 2919 | ||
| GenBank mRNA | AF189285.1 | - | 11 | 2919 | ||
| BC026792.1 | - | 11 | 2710 | 4911382 | ||
| BC051036.1 | - | 10 | 2738 | |||