siRNA Details
| Gene Symbol: Tmprss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015775.2 | NP_056590.2 | 5 | 655 | ||
| GenBank mRNA | AF113596.1 | - | 5 | 525 | ||
| AF199362.1 | - | 5 | 536 | |||
| AF243500.1 | - | 5 | 525 | |||
| AK141350.1 | - | 5 | 630 | |||
| AK146047.1 | - | 5 | 539 | |||
| AK161173.1 | - | 5 | 649 | |||
| BC038393.1 | - | 5 | 494 | 2648878 | ||
| BC054348.1 | - | 5 | 655 | 2648889 | ||

Tmprss2
Genomic Map