siRNA Details
| Gene Symbol: Postn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198765.1 | NP_001185694.1 | 6 | 688 | ||
| NM_001198766.1 | NP_001185695.1 | 6 | 688 | |||
| NM_015784.3 | NP_056599.1 | 6 | 688 | |||
| GenBank mRNA | AK030756.1 | - | 6 | 687 | ||
| AK154049.1 | - | 6 | 686 | |||
| AY651928.1 | - | 6 | 658 | |||
| BC007141.1 | - | 6 | 682 | |||
| BC031449.1 | - | 6 | 657 | 4457222 | ||
| D13664.1 | - | 6 | 670 | |||
| E05655.1 | - | 6 | 684 | |||
| KM117171.1 | - | 6 | 652 | |||
| KM117172.1 | - | 6 | 652 | |||

Postn
Genomic Map