siRNA Details
| Gene Symbol: Fbxw7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177773.1 | NP_001171244.1 | 13 | 2101 | ||
| NM_001177774.1 | NP_001171245.1 | 12 | 2010 | |||
| NM_080428.3 | NP_536353.2 | 10 | 1646 | |||
| GenBank mRNA | AB221593.1 | - | 10 | 1488 | ||
| AF391192.1 | - | 10 | 1646 | |||
| AF427101.1 | - | 10 | 1568 | |||
| AK032174.1 | - | 4 | 572 | |||
| BC060620.1 | - | 13 | 2101 | |||
| BC066093.1 | - | 12 | 2011 | |||
| BC085184.1 | - | 12 | 3802 | 30664813 | ||
| BC131648.1 | - | 10 | 1510 | 40110349 | ||

Fbxw7
Genomic Map