siRNA Details
| Gene Symbol: Fbxo6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163704.1 | NP_001157176.1 | 6 | 1138 | ||
| NM_001163705.1 | NP_001157177.1 | 6 | 922 | |||
| NM_001163706.1 | NP_001157178.1 | 6 | 914 | |||
| NM_001163707.1 | NP_001157179.1 | 6 | 949 | |||
| NM_015797.4 | NP_056612.1 | 6 | 1664 | |||
| GenBank mRNA | AF176526.1 | - | 6 | 1037 | ||
| AK002840.1 | - | 6 | 876 | |||
| AK144715.1 | - | 6 | 1075 | |||
| AK150612.1 | - | 6 | 1106 | |||
| AK152888.1 | - | 6 | 988 | |||
| AK165867.1 | - | 6 | 1649 | |||
| AK170743.1 | - | 6 | 915 | |||
| AK171483.1 | - | 6 | 915 | |||
| BC017512.1 | - | 6 | 934 | 4485218 | ||

Fbxo6
Genomic Map