siRNA Details
Gene Symbol: Crim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015800.3 | NP_056615.1 | 13 | 2762 | ||
GenBank mRNA | AF168680.1 | - | 13 | 2214 | ||
AK053197.1 | - | 11 | 1705 | |||
BC100348.1 | - | 13 | 2547 | 30521148 |
