siRNA Details
| Gene Symbol: Mapk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015806.5 | NP_056621.4 | 6 | 2403 | ||
| NM_027418.2 | NP_081694.1 | 6 | 2407 | |||
| GenBank mRNA | AF132850.1 | - | 5 | 2136 | ||
| BC006778.1 | - | 5 | 1617 | |||
| BC024684.1 | - | 2 | 702 | |||
| BC037060.1 | - | 5 | 2141 | |||
| BC048779.1 | - | 5 | 1905 | 6315375 | ||
| BC052420.1 | - | 5 | 1652 | 5702905 | ||
| BC100385.1 | - | 6 | 2385 | 30732053 | ||

Mapk6
Genomic Map