siRNA Details
| Gene Symbol: Rgs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081650.2 | NP_001075119.1 | 6 | 713 | ||
| NM_019492.3 | NP_062365.2 | 15 | 1417 | |||
| NM_134257.3 | NP_599018.3 | 6 | 713 | |||
| GenBank mRNA | AF319519.1 | - | 6 | 726 | ||
| AF350047.1 | - | 6 | 696 | |||
| AJ250999.1 | - | 15 | 1417 | |||
| AK015722.1 | - | 13 | 1172 | |||
| AK028445.1 | - | 6 | 698 | |||

Rgs3
Genomic Map