siRNA Details
| Gene Symbol: Rgs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081650.2 | NP_001075119.1 | 8 | 1070 | ||
| NM_019492.3 | NP_062365.2 | 17 | 1774 | |||
| NM_134257.3 | NP_599018.3 | 8 | 1070 | |||
| GenBank mRNA | AF319519.1 | - | 8 | 1083 | ||
| AF350047.1 | - | 8 | 1053 | |||
| AJ250999.1 | - | 17 | 1774 | |||
| AK004648.1 | - | 2 | 239 | |||
| AK015722.1 | - | 15 | 1531 | |||
| AK028445.1 | - | 8 | 1055 | |||
| AK041392.1 | - | 2 | 246 | |||
| BY704855.1 | - | 2 | 234 | |||

Rgs3
Genomic Map