siRNA Details
| Gene Symbol: Dkk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015814.2 | NP_056629.1 | 4 | 484 | ||
| GenBank mRNA | AF177400.1 | - | 4 | 471 | ||
| AJ243964.1 | - | 4 | 487 | |||
| AK004853.1 | - | 4 | 484 | |||
| AK013054.2 | - | 3 | 422 | |||
| AK013622.1 | - | 3 | 396 | |||
| BC046304.1 | - | 3 | 399 | 6493718 | ||
| BC050934.1 | - | 3 | 381 | 6812621 | ||

Dkk3
Genomic Map