siRNA Details
Gene Symbol: Magi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170745.1 | NP_001164216.1 | 17, 18 | 3426 | ||
NM_001170746.1 | NP_001164217.1 | 19, 20 | 3633 | |||
NM_015823.3 | NP_056638.1 | 18, 19 | 3468 | |||
GenBank mRNA | AK039336.1 | - | 15, 16 | 2453 | ||
AK122349.1 | - | 17, 18 | 2928 | |||
AK147530.1 | - | 17, 18 | 3427 | |||
BC059005.1 | - | 18, 19 | 3125 | 6847238 |
