siRNA Details
| Gene Symbol: Gne | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190414.1 | NP_001177343.1 | 2, 3 | 506 | ||
| NM_015828.3 | NP_056643.3 | 2, 3 | 324 | |||
| GenBank mRNA | AK136573.1 | - | 2, 3 | 325 | ||
| AK170707.1 | - | 2, 3 | 507 | |||
| AK199019.1 | - | 1, 2 | 27 | |||
| AM697710.1 | - | 2, 3 | 245 | |||
| BC015277.1 | - | 2, 3 | 312 | 4216299 | ||
| BC051254.1 | - | 2, 3 | 312 | 6478532 | ||

Gne
Genomic Map