siRNA Details
| Gene Symbol: Gne | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190414.1 | NP_001177343.1 | 7 | 1451 | ||
| NM_015828.3 | NP_056643.3 | 7 | 1269 | |||
| GenBank mRNA | AJ132236.1 | - | 6 | 1097 | ||
| AK010070.1 | - | 4 | 417 | |||
| AK136573.1 | - | 7 | 1270 | |||
| AK170707.1 | - | 7 | 1452 | |||
| AM697710.1 | - | 7 | 1190 | |||
| BC015277.1 | - | 7 | 1257 | 4216299 | ||
| BC051254.1 | - | 7 | 1257 | 6478532 | ||

Gne
Genomic Map