siRNA Details
| Gene Symbol: Chek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016681.3 | NP_057890.1 | 5 | 698 | ||
| GenBank mRNA | AF086905.1 | - | 4 | 659 | ||
| AK044913.1 | - | 5 | 699 | |||
| BC056617.1 | - | 5 | 722 | 4977960 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chek2| Gene Symbol: Chek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016681.3 | NP_057890.1 | 5 | 698 | ||
| GenBank mRNA | AF086905.1 | - | 4 | 659 | ||
| AK044913.1 | - | 5 | 699 | |||
| BC056617.1 | - | 5 | 722 | 4977960 | ||